missing translation for 'onlineSavingsMsg'
Learn More

Phenazine 99.0+%, TCI America™

Catalog No. P00825G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00825G 5 g
P00821G 1 g
P008225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P00825G Supplier TCI America Supplier No. P00825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-82-0
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00005023
InChI Key PCNDJXKNXGMECE-UHFFFAOYSA-N
Synonym dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
PubChem CID 4757
ChEBI CHEBI:36674
IUPAC Name phenazine
SMILES C1=CC=C2C(=C1)N=C3C=CC=CC3=N2

Specifications

Melting Point 176°C
Color Yellow
Quantity 5 g
Formula Weight 180.21
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenazine
RTECSNumber SG1360000
TSCA Yes