missing translation for 'onlineSavingsMsg'
Learn More

Phenazine Methyl Sulfate 98.0+%, TCI America™

Catalog No. P00831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00831G 1 g
P00835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00831G Supplier TCI America Supplier No. P00831G

Chemical Identifiers

CAS 299-11-6
Molecular Formula C14H14N2O4S
Molecular Weight (g/mol) 306.336
MDL Number MFCD00011923
InChI Key RXGJTUSBYWCRBK-UHFFFAOYSA-M
Synonym phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
PubChem CID 9285
ChEBI CHEBI:8055
IUPAC Name 5-methylphenazin-5-ium;methyl sulfate
SMILES C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]

Specifications

Melting Point 160°C
Color Green-Yellow
Quantity 1 g
Formula Weight 306.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Phenazine Methyl Sulfate
RTECSNumber SG1645000
TSCA Yes