missing translation for 'onlineSavingsMsg'
Learn More

Phenazine methosulfate, 98+%

Catalog No. p-7037879
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5671806 5 g
AAH5671814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5671806 Supplier Thermo Scientific Chemicals Supplier No. H5671806
Only null left

CAS: 299-11-6 | C14H14N2O4S | 306.336 g/mol

It is used with ascorbic acid to determine nitric oxide reductase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used with ascorbic acid to determine nitric oxide reductase activity.

Solubility
Soluble in water. (100g/L at20°C)

Notes
Light sensitive. Incompatible with oxidizing agents. Store at -20°C.

Chemical Identifiers

CAS 299-11-6
Molecular Formula C14H14N2O4S
Molecular Weight (g/mol) 306.336
MDL Number MFCD00011923
InChI Key RXGJTUSBYWCRBK-UHFFFAOYSA-M
Synonym phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
PubChem CID 9285
ChEBI CHEBI:8055
IUPAC Name 5-methylphenazin-5-ium;methyl sulfate
SMILES C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]

Specifications

Quantity 5 g
Beilstein 3898869
Sensitivity Light sensitive
Solubility Information Soluble in water. (100g/L at20°C)
Formula Weight 306.34
Percent Purity ≥98%
Chemical Name or Material Phenazine methosulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-072-1
RTECSNumber SG1645000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only