missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenazine, 99+%
Description
Phenazine is the parent substance of many dyestuffs, such as the toluylene red, indulines and safranines. It can be applied in therapeutic uses, used as antipsychotic and antiemetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 175°C to 177°C |
| Quantity | 5 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 180.21 |
| Percent Purity | ≥99% |
| Chemical Name or Material | Phenazine |
Safety and Handling
| Hazard Category | H302-H341 |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| EINECSNumber | 202-193-9 |
| RTECSNumber | SG1360000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only