missing translation for 'onlineSavingsMsg'
Learn More

Phenanthridine, 98%

Catalog No. AC169250050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169250050 5 g Glass bottle
AC169250010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169250050 Supplier Thermo Scientific Chemicals Supplier No. 169250050
Only null left

CAS: 229-87-8 | C13H9N | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 229-87-8
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00004989
InChI Key RDOWQLZANAYVLL-UHFFFAOYSA-N
Synonym 6-phenanthridine, benzo c quinoline, 3,4-benzoquinoline, 9-azaphenanthrene, 3,4-benzoisoquinoline, 5-azaphenanthrene, unii-62qgs7cps6, ccris 1234, 62qgs7cps6
PubChem CID 9189
ChEBI CHEBI:36421
IUPAC Name phenanthridine
SMILES C1=CC=C2C(=C1)C=NC3=CC=CC=C23

Specifications

Melting Point 104.0°C to 108.0°C
Color White to Beige
Density 100g/mL
Boiling Point 349.0°C (769.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 20, 466
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene, dmso
Formula Weight 179.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenanthridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-934-4

RUO – Research Use Only