Learn More
Phenanthrene, tech. 90%
CAS: 85-01-8 | C14H10 | 178.23 g/mol
Supplier: Thermo Scientific Chemicals A1964622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsPhenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
Solubility
Soluble in alcohol, benzene, toluene, and glacial acetic acid
Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
Specifications
| Phenanthrene | |
| 97°C to 101°C | |
| 340°C | |
| C14H10 | |
| 100 g | |
| 1905428 | |
| phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730 | |
| YNPNZTXNASCQKK-UHFFFAOYSA-N | |
| phenanthrene | |
| 995 | |
| 178.23 | |
| Technical |
| 85-01-8 | |
| 0.98 | |
| 171°C (339°F) | |
| MFCD00001168 | |
| UN3077 | |
| 14,7212 | |
| Soluble in alcohol, benzene, toluene, and glacial acetic acid | |
| C1=CC=C2C(C=CC3=CC=CC=C23)=C1 | |
| 178.23 | |
| CHEBI:28851 | |
| 90% |
Chemical Identifiers
| 85-01-8 | |
| 178.23 | |
| YNPNZTXNASCQKK-UHFFFAOYSA-N | |
| 995 | |
| phenanthrene |
| C14H10 | |
| MFCD00001168 | |
| phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730 | |
| CHEBI:28851 | |
| C1=CC=C2C(C=CC3=CC=CC=C23)=C1 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 201-581-5
RTECSNumber : SF7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only