missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, tech. 90%

CAS: 85-01-8 | C14H10 | 178.23 g/mol

Supplier:  Thermo Scientific Chemicals A196460E

 View more versions of this product

Catalog No. AAA196460E


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

Solubility
Soluble in alcohol, benzene, toluene, and glacial acetic acid

Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
Specifications

Specifications

Phenanthrene
97°C to 101°C
340°C
C14H10
2500 g
1905428
phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
YNPNZTXNASCQKK-UHFFFAOYSA-N
phenanthrene
995
178.23
Technical
85-01-8
0.98
171°C (339°F)
MFCD00001168
UN3077
14,7212
Soluble in alcohol, benzene, toluene, and glacial acetic acid
C1=CC=C2C(C=CC3=CC=CC=C23)=C1
178.23
CHEBI:28851
90%

Chemical Identifiers

85-01-8
178.23
YNPNZTXNASCQKK-UHFFFAOYSA-N
995
phenanthrene
C14H10
MFCD00001168
phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
CHEBI:28851
C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 201-581-5

RTECSNumber : SF7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only