missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, 97%

Catalog No. p-180791
Change view
Click to view available options
Quantity:
5 g
50 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130090050 5 g Glass bottle
AC130090500 50 g Plastic bottle
AC130095000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130090050 Supplier Thermo Scientific Chemicals Supplier No. 130090050
Only null left

CAS: 85-01-8 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 95°C to 101°C
Color White to Gray
Density 1.0630g/mL
Boiling Point 336°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05, 667
Specific Gravity 1.063
Merck Index 15, 7326
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetic acid,soluble in alcohol,ether,benzene and carbon,disulfide
Formula Weight 178.23
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenanthrene, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 201-581-5
RTECSNumber SF7175000
TSCA TSCA

RUO – Research Use Only