missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, 90%, Spectrum™ Chemical

Catalog No. 18612214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612214 25 g Poly Bottle
18612211 100 g Poly Bottle
18612215 500 g Poly Bottle
18612213 2.5 kg Poly Pail
18612212 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612214 Supplier Spectrum Chemical Mfg Cor Supplier No. PH10625GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 85-01-8

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 25 g
Percent Purity 0.9
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.