missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-9-carboxaldehyde, 97%

Catalog No. p-7028369
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2564803 1 g
AAB2564806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2564803 Supplier Thermo Scientific Chemicals Supplier No. B2564803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4707-71-5 | C15H10O | 206.24 g/mol

Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

Solubility
Soluble in acetone, dichloromethane, methanol.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.

Chemical Identifiers

CAS 4707-71-5
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001175
InChI Key QECIGCMPORCORE-UHFFFAOYSA-N
Synonym 9-phenanthrenecarboxaldehyde, phenanthrene-9-carboxaldehyde, 9-formylphenanthrene, phenanthrene-9-aldehyde, 9-phenanthrenecarbaldehyde, 9-phenanthrenealdehyde, 9-phenanthrencarbaldehyd, 9-phenanthraldehyde, 9-phenanthrenaldehyde, phenanthren-9-aldehyd
PubChem CID 78437
IUPAC Name phenanthrene-9-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 100°C to 103°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in acetone,dichloromethane,methanol.
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material Phenanthrene-9-carboxaldehyde
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 225-194-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only