missing translation for 'onlineSavingsMsg'
Learn More

Phenacyltriphenylphosphonium Bromide 97.0+%, TCI America™

Catalog No. P118225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P118225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P118225G Supplier TCI America Supplier No. P118225G
Only null left

Chemical Identifiers

CAS 6048-29-9
Molecular Formula C26H22BrOP
Molecular Weight (g/mol) 461.34
MDL Number MFCD00011920
InChI Key AEHDSYHVTDJGDN-UHFFFAOYSA-M
Synonym phenacyltriphenylphosphonium bromide, 2-oxo-2-phenylethyl triphenylphosphonium bromide, phenacyltriphenylphosphoniumbromide, phosphonium, 2-oxo-2-phenylethyl triphenyl-, bromide, phenacyl triphenyl phosphanium bromide, 2-oxo-2-phenylethyl triphenylphosphanium bromide, acmc-209mk8, wln: rv1pr&r&r &e, phenacyl triphenyl phosphonium bromide, phosphonium, 2-oxo-2-phenylethyl triphenyl-, bromide 1:1
PubChem CID 197064
IUPAC Name (2-oxo-2-phenylethyl)triphenylphosphanium bromide
SMILES [Br-].O=C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 268°C
Quantity 25 g
Formula Weight 461.34
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenacyltriphenylphosphonium Bromide
RTECSNumber TA2400000
TSCA No