missing translation for 'onlineSavingsMsg'
Learn More

Phenacyl Bromide 98.0+%, TCI America™

Catalog No. p-7118504
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B053525G 25 g
B0535100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B053525G Supplier TCI America Supplier No. B053525G
Only null left

Chemical Identifiers

CAS 70-11-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD00000195
InChI Key LIGACIXOYTUXAW-UHFFFAOYSA-N
Synonym 2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
PubChem CID 6259
ChEBI CHEBI:51846
IUPAC Name 2-bromo-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 135°C
Quantity 25 g
UN Number 2645
Formula Weight 199.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenacyl Bromide
EINECSNumber (3)-2931
TSCA Yes
Recommended Storage Refrigerator