missing translation for 'onlineSavingsMsg'
Learn More

Phenacyl 4-(Bromomethyl)phenylacetate 97.0+%, TCI America™

Catalog No. B13151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13151G 1 g
B13155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13151G Supplier TCI America Supplier No. B13151G
Only null left

Chemical Identifiers

CAS 66270-97-1
Molecular Formula C17H15BrO3
Molecular Weight (g/mol) 347.21
MDL Number MFCD00009632
InChI Key BUQPIKAEKYNDAS-UHFFFAOYSA-N
Synonym 4-(Bromomethyl)phenylacetic Acid Phenacyl Ester
PubChem CID 570776
IUPAC Name 2-oxo-2-phenylethyl 2-[4-(bromomethyl)phenyl]acetate
SMILES BrCC1=CC=C(CC(=O)OCC(=O)C2=CC=CC=C2)C=C1

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 1 g
Formula Weight 347.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenacyl 4-(Bromomethyl)phenylacetate