missing translation for 'onlineSavingsMsg'
Learn More

Phenaceturic Acid 98.0+%, TCI America™

Supplier:  TCI America P013125G

Catalog No. P013125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenaceturic Acid
144°C
C10H11NO3
25 g
UTYVDVLMYQPLQB-UHFFFAOYSA-N
2-(2-phenylacetamido)acetic acid
68144
193.20
Crystalline Powder
500-98-1
White
MFCD00021744
N-(Phenylacetyl)glycine
OC(=O)CNC(=O)CC1=CC=CC=C1
193.20
CHEBI:27480
≥98.0% (HPLC,T)

Chemical Identifiers

500-98-1
193.20
UTYVDVLMYQPLQB-UHFFFAOYSA-N
68144
2-(2-phenylacetamido)acetic acid
C10H11NO3
MFCD00021744
N-(Phenylacetyl)glycine
CHEBI:27480
OC(=O)CNC(=O)CC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions