missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PHARM-TROL™ Count Precision Size Standards

Document the reproducibility of a particle counter by permitting a continuous record of its performance.

$1,584.94 - $3,965.78

Specifications

Concentration 3800 particles/mL ± 15%
Contents Polystyrene microspheres in water
Color White or Colorless
For Use With (Application) USP Verification
Material Polystyrene
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity & Availability  
09980236
View Documents Promotion Details
Thermo Scientific™
CSBX
15 μm 20 x 25 mL Bottles
Pack of 20 Each for $3,965.78
Only null left
 
09980235
View Documents Promotion Details
Thermo Scientific™
CSPK
15.0 ±0.2 μm 6 x 25 mL Bottles
Pack of 6 Each for $1,584.94
Only null left
 
Description

Description

Document the reproducibility of a particle counter by permitting a continuous record of its performance with Thermo Scientific™ PHARM-TROL™ Count Precision Size Standards, ready-to-use precision standards designed for regular use in liquid particle counters for USP <788> and <789> verification. Data is documented for internal or customer audits by using a NIST™-traceable microsphere suspension with a known concentration.

Features:

  • Contains NIST™-traceable size standards with a measured particle count
  • Developed for manufacturers of parenteral drugs and ophthalmic solutions seeking interim verification of USP <788> and <789> verification (Particulate Matter in Injections and Particulate Matter in Opthalmic Solutions)
  • Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology
  • Prepared with the same exacting procedures as the USP Count Reference Standard, providing process control data for particle counting consistency and verification of size calibration
Specifications

Specifications

3800 particles/mL ± 15%
White or Colorless
Polystyrene
Certificate of Calibration and Traceability to NIST
1.59 at 589nm (25°C)
Product stable for a minimum of 12 months
Polystyrene microspheres in water
USP Verification
Count Control Particles
1.05g/cm3
<3%
Videos
SDS
Documents

Documents