Learn More
Promega pHAb Thiol Reactive Dye
pHAb Amine Reactive Dye exhibits very low fluorescence at pH > 7 and much higher fluorescence as the pH of the solution becomes acidic. pHAb Dyes are designed specifically for antibody labeling.
Supplier:  Promega G9831
Encompass
 View more versions of this product
Catalog No. PRG9831
Only null left
Description
Description
  • Measure Antibody Internalization in Real Time
  • Compatible with 96-well, plate-based assay
  • High signal-to-background ratios
  • On-Bead conjugation
Specifications
Chemical Identifiers
-30° to -10°C for 1 month and <-65°C for long-term storage
Specifications
250 μg
-30° to -10°C for 1 month and <-65°C for long-term storage
Product Suggestions
Product Suggestions