Learn More
Promega pHAb Amine Reactive Dye
Catalog No. PRG9845
Encompass
Change view
Click to view available options
Quantity:
4 x 250 μg
Catalog No. Quantity
PRG9845 4 x 250 μg
1 options
Catalog No. PRG9845 Supplier Promega Supplier No. G9845
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
pHAb Amine Reactive Dye exhibits very low fluorescence at pH > 7 and much higher fluorescence as the pH of the solution becomes acidic. pHAb Dyes are designed specifically for antibody labeling.
  • Measure Antibody Internalization in Real Time
  • Compatible with 96-well, plate-based assay
  • High signal-to-background ratios
  • On-Bead conjugation
Specifications
Quantity 4 x 250 μg
Content And Storage <-65°C