missing translation for 'onlineSavingsMsg'
Learn More

PHA Reagent Grade

Catalog No. R30852701
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30852701 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R30852701 Supplier Thermo Fisher Scientific Supplier No. RE30852701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Facilitate cytogenic studies of chromosomes by stimulating the mitotic division of lymphocytes with this reagent.

Facilitate cytogenic studies of chromosomes by stimulating the mitotic division of lymphocytes maintained in a cell culture system with Thermo Scientific™ Remel™ PHA Reagent Grade. Phytohaemagglutinin (PHA) is universally effective and convenient stimulant of lymphocytes from many animal species. The widespread popularity of peripheral blood culture as a means of chromosome analysis has been largely dependent on this property.

Use Remel Phytohaemagglutinin to stimulate mitotic division of lymphocytes maintained in a cell culture system and facilitate cytogenic studies of chromosomes.

Specifications

Content And Storage 2°C to 8°C
Description Phytohaemagglutinin (Reagent Grade)
CE Marker Yes
Type PHA
Includes 1 bottle Phytohaemagglutinins (reagent grade)
Quantity 5 mL
Species Animal