missing translation for 'onlineSavingsMsg'
Learn More

YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System

Measure pH and pH & ORP/Redox quickly and easily

Supplier:  YSI™ 5565

 View more versions of this product

Catalog No. 13-298-66


Only null left
Explore available promotions

Description

Description

  • User-replaceble sensor allows for quick sensor replacements if needed
  • No tools necessary

Specifications

Specifications

pH and pH/ORP probe
Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications
YSI model 556 handheld multi probe meter
Storage bottle with storage solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions