missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ pH Meter SevenDirect SD20

The SevenDirect SD20 is an accurate and flexible benchtop meter for the determination of pH and ORP. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

Supplier:  Mettler Toledo™ 30671559

 View more versions of this product

Catalog No. 01911917


Earn Up to $2,000 in Accessories with Purchase
View all promotions
Only null left

Description

Description

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
Specifications

Chemical Identifiers

0.002
PC, Printer, USB Stick
0.001, 0.01, 0.1
Up to 5
-22°F to +266°F
Color TFT
2000 Standard Measurements
Automatic, Manual
195 x 205 x 65 mm
1.9 lb.
±0.1°C between 0°C and 100°C
-2 to 20
-2000 to 2000 mV
-30°C to +130°C
0.1°C
Yes
2000
IP54
850 g

Specifications

Instrument, power s upply, protective cover, Get Started guide, electrode holder EasyPlace, appropriate electrode, calibration solutions
±0.1°C between 0°C and 100°C
PC, Printer, USB Stick
Linear/Segmented
0.001, 0.01, 0.1
1, 0.1 mV
-30°C to +130°C
0.1°C
Yes
2000
IP54
850 g
1
SevenDirect SD20 Solids Kit
0.1 mV
0.002
pH with mV and Temperature
-2 to 20
-2000 to 2000 mV
Up to 5
-22°F to +266°F
Color TFT
2000 Standard Measurements
Automatic, Manual
195 x 205 x 65 mm
1.9 lb.
InLab Solids-Pro ISM pH/Temperature Sensor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions