missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ pH Meter SevenDirect SD20
The SevenDirect SD20 is an accurate and flexible benchtop meter for the determination of pH and ORP. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.
Supplier: Mettler Toledo™ 30671556
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH and ORP of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.
Chemical Identifiers
| 0.002 | |
| PC, Printer, USB Stick | |
| 0.001, 0.01, 0.1 | |
| Up to 5 | |
| -22°F to +266°F | |
| Color TFT | |
| 2000 Standard Measurements | |
| Automatic, Manual | |
| 195 x 205 x 65 mm | |
| 1.9 lb. |
| ±0.1°C between 0°C and 100°C | |
| -2 to 20 | |
| -2000 to 2000 mV | |
| -30°C to +130°C | |
| 0.1°C | |
| Yes | |
| 2000 | |
| IP54 | |
| 850 g |
Specifications
| Instrument, power s upply, protective cover, Get Started guide, electrode holder EasyPlace, appropriate electrode, calibration solutions, EasyMix magnatic stirrer | |
| 0.002 mV | |
| PC, Printer, USB Stick | |
| Linear/Segmented | |
| 0.001, 0.01, 0.1 | |
| 0.1, 0.01 mV | |
| -30°C to +130°C | |
| 0.1°C | |
| Yes | |
| 2000 | |
| IP54 | |
| 850 g | |
| 2 | |
| SevenDirect SD20 HA Kit |
| 0.002 | |
| ±0.1°C between 0°C and 100°C | |
| pH with mV and Temperature | |
| -2 to 20 | |
| -2000 to 2000 mV | |
| Up to 5 | |
| -22°F to +266°F | |
| Color TFT | |
| 2000 Standard Measurements | |
| Automatic, Manual | |
| 195 x 205 x 65 mm | |
| 1.9 lb. | |
| InLab Routine-Pro ISM pH/Temperature Sensor |