Learn More
New England Biolab Ltd Ph.D.™ Peptide Display Cloning System 20 g
Catalog No. 50811592
Encompass
The Ph.D. Peptide Display Cloning System contains M13KE vector, the parent vector of NEB's Ph.D. libraries. The included M13 extension primer is used to make DNA duplex out of a user provided synthetic oligo, with or without random bases. The display system is best used to display small peptides, i.e. sequences of 50 amino acids or less.
Catalog No. 50811592 Supplier New England Biolab Ltd Supplier No. E8101S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Ph.D. Peptide Display Cloning System contains M13KE vector, the parent vector of NEB's Ph.D. libraries. The included M13 extension primer is used to make DNA duplex out of a user provided synthetic oligo, with or without random bases. The display system is best used to display small peptides, i.e. sequences of 50 amino acids or less.