missing translation for 'onlineSavingsMsg'
Learn More

pH 6 Buffer Standard, SPEX CertiPrep™

Catalog No. PHBUFF6500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHBUFF6500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHBUFF6500 Supplier SPEX CertiPrep Group LLC Supplier No. PHBUFF6500
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
pH6 SI Units

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material pH 6 Buffer Standard
Concentration 6 mol/L
For Use With (Application) pH Calibration
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Physical Form Liquid
Quantity 500 mL
Solution Type Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition (by Analyte or Components) Various

Product intended for laboratory use only