missing translation for 'onlineSavingsMsg'
Learn More

Verispec™ PFAS Standards, Certified Reference Material

Catalog No. 01341381
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01341381 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01341381 Supplier Verispec™ Supplier No. RV020003100CM

Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.

  • Available in convenient 100 mg quantities for customizable calibration standards
  • Supports environmental testing, drinking and wastewater analysis, food safety, and industrial quality control.
  • Ensure compliance with key methods:

EPA: 533, 537.1, 1633, 8327

ASTM: D7968, D7979, D8421, D8535

  • High-quality sample preparation solutions, solvents, and mobile phase reagents
  • Minimize contamination risks to enhance accuracy in sensitive applications
  • Contact Us for customs or ready to use standards

Specifications

Chemical Name or Material Perfluorotetradecanoic acid
CAS 376-06-7
Quantity 100 mg
Molecular Weight (g/mol) 714.11 g/mol
Percent Purity Lot Specific
Physical Form Solid or Liquid