missing translation for 'onlineSavingsMsg'
Learn More

Verispec™ PFAS Standards, Certified Reference Material

Catalog No. p-200102288
Change view
missing translation for 'orderingAttributeHoverText'
Chemical Name or Material:
N-Ethyl-N-(2-hydroxyethyl)-PFOS
PFOS tetraethylammonium salt
Perfluorooctane sulfonic acid
Perfluorooctanesulphonamide
Potassium nonafluoro-1-butanesulfonate
Molecular Weight (g/mol):
338.18 g/mol
499.14 g/mol
500.12 g/mol
571.25 g/mol
629.37 g/mol
Catalog No. Chemical Name or Material Molecular Weight (g/mol)
01341375 PFOS tetraethylammonium salt 629.37 g/mol
01341377 N-Ethyl-N-(2-hydroxyethyl)-PFOS 571.25 g/mol
01341378 Perfluorooctanesulphonamide 499.14 g/mol
01341379 Perfluorooctane sulfonic acid 500.12 g/mol
01341384 Potassium nonafluoro-1-butanesulfonate 338.18 g/mol
5 options

Catalog No. 01341375

Supplier: Verispec™ RV020009100CM

Only null left

Manufactured and tested in an ISO 17025 and 17034 accredited facility for applications in environmental testing, food safety, water quality, and more.

  • Available in convenient 100 mg quantities for customizable calibration standards
  • Supports environmental testing, drinking and wastewater analysis, food safety, and industrial quality control.
  • Ensure compliance with key methods:

EPA: 533, 537.1, 1633, 8327

ASTM: D7968, D7979, D8421, D8535

  • High-quality sample preparation solutions, solvents, and mobile phase reagents
  • Minimize contamination risks to enhance accuracy in sensitive applications
  • Contact Us for customs or ready to use standards

Specifications

Chemical Name or Material PFOS tetraethylammonium salt
CAS 56773-42-3
Quantity 100 mg
Molecular Weight (g/mol) 629.37 g/mol
Percent Purity Lot Specific
Physical Form Solid or Liquid