Learn More
Petroleum ether, extra pure, boiling range 60-95°C
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Supplier: Thermo Scientific Chemicals 292510025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Petroleum ether | |
| 64742-49-0 | |
| 60.0°C to 95.0°C | |
| −26°C | |
| Glass Bottle | |
| 1.3880 to 1.3920 (20°C, 589nm) | |
| 0.7 | |
| MFCD00081849 | |
| isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
| CCCC(C)C | |
| 86.178 | |
| Extra Pure |
| -40.0°C | |
| 0.7000g/mL | |
| 2.5 L | |
| Authentic | |
| Liquid | |
| 0.005% max. | |
| C6H14 | |
| 0.57 mm2/s (25°C) | |
| AFABGHUZZDYHJO-UHFFFAOYSA-N | |
| Petroleum ether | |
| 7892 |
Chemical Identifiers
| 64742-49-0 | |
| 86.178 | |
| AFABGHUZZDYHJO-UHFFFAOYSA-N | |
| 7892 |
| C6H14 | |
| MFCD00081849 | |
| isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
| CCCC(C)C |
Safety and Handling
GHS P Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only