missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, ACS Reagent (Ligroin), Reagents

Catalog No. p-200021171
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
Tin Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337999 1 L Glass Bottle
01337998 200 L Steel Drum
01337997 20 L Steel Pail
01337996 4 L Glass Bottle
01337994 10 L Tin Can
01337993 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337999 Supplier Labchem Inc Supplier No. C11990001C
Only null left

Petroleum Ether, ACS Reagent (Ligroin), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Petroleum Ether, ACS Reagent (Ligroin)
CAS 8032-32-4
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.645
Molecular Formula C6-16H12-34
Synonym Naphtha, Petroleum benzine, Petroleum spirit, Ligroin, Ligroine
Boiling Point 35°C
Grade ACS
Show More Show Less