Learn More
George Taylor Brass & Bronze Works Petroleum Bacon Bomb Sampler
For composite sampling by ASTM™ D 4057-95
Supplier:  George Taylor Brass & Bronze Works TBB 4850
Encompass
 View more versions of this product
Catalog No. 14209
Only null left
Description
Description
For composite sampling by ASTM™ D 4057-95
  • Weighted sampler obtains samples of any liquid
  • Automatically opens when plunger touches bottom
  • Manually opens at desired depth with a pull cord or extension rod
  • Sampler closes when lifted and can be locked for transport
  • No O-rings to wear out or lose
  • Antimagnetic
  • Brass and bronze construction with heavy nickel plating
  • Plunger threaded 1/4-20 to accept threaded extension rods
Specifications
Chemical Identifiers
Petroleum Bacon Bomb Sampler
Threaded plunger
ASTM D4057-95
Brass, Bronze
Heavy Nickel
Ideal for obtaining samples at various levels of any liquid
Specifications
Petroleum Bacon Bomb Sampler
Threaded plunger
500 mL
ASTM D4057-95
2.75 in.
12 in.
Brass, Bronze
16 oz.
Heavy Nickel
Ideal for obtaining samples at various levels of any liquid
6.9 cm
30.4 cm
Product Suggestions
Product Suggestions