Learn More
Foxx Life Sciences PETG MEDIA BOTTLE 500ML STERIL
Supplier:  Foxx Life Sciences 1177RLS
Encompass
1177-RLS: Media Bottle, 500 mL, PETG, Sterile. Foxx Life Sciences' PETG Media Bottles have been designed specifically for life science researchers to store common solutions such as tissue culture media, serum, and buffers. Our PETG bottles are optically clear, and offer better impact resistance than polystyrene storage bottles. The bottles are gas-impermeable, and are an ideal low-cost choice for general laboratory solution storage. They are also designed to handle full vacuum, and can be used with the Autofil filtration system. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying of the bottle. The innovative cap also allows for easy uncapping, tightening, and bottle stacking and provides a large surface for labeling.
Catalog No. 501050041
Only null left