missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ PET Liquid Scintillation Vials

Catalog No. 0334171E
Change view
Click to view available options
Material (Liner):
Foamed Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner)
0334171E Foamed Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0334171E Supplier Wheaton™ Supplier No. 986750

For optimal performance in the laboratory. Wheaton™ PET Liquid Scintillation Vials offer low permeability to solvents and minimal background counts.

  • With caps attached
  • Available linerless or with foamed polyethylene liner

Specifications

Material (Liner) Foamed Polyethylene
Diameter (Metric) Outer 27 mm
Material PET
Volume (Metric) 20 mL
Closure Material Polypropylene
Closure Type Screw Cap
Quantity 500 Pk.
Closure Size 22-400
Height (Metric) 61 mm
Product Type Scintillation Vial