missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Triazoxid, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 72459-58-6 |
| Physical Form | Neat |
| Linear Formula | C10H6 ClN5 O |
| MDL Number | MFCD00145438 |
| Melting Point | 178°C to 182°C |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 247.64 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH