missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Tembotrione, MilliporeSigma™ Supelco™

Catalog No. 111019128
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019128 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019128 Supplier Merck Emd Millipore Supplier No. 32766100MG
Only null left

Tembotrione is a member of β-triketone family of herbicides that is utilized for grass and broadleaf weed control during early to mid-post-emergence of field corn.

Agriculture

Specifications

Product Line PESTANAL
CAS 335104-84-2
Physical Form Neat
Linear Formula C17H16 ClF3O6 S
MDL Number MFCD11840613
Synonym 2-{2-Chloro-4-(methylsulfonyl)-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl}-1,3-cyclohexanedione; 2-{2-Chloro-4-mesyl-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl}cyclohexane-1,3-dione
Type Analytical Standard
Quantity 100 mg
Formula Weight 440.82
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH