missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ (S)-3-Anilino-5-methyl-5-phenylimidazolidine-2,4-dione Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022461
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022461 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022461 Supplier Merck Emd Millipore Supplier No. 34183100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 332855-88-6
Flash Point Not Applicable
Molecular Formula C16H15N3O2
Linear Formula C16H15N3O2
Quantity 100 mg
MDL Number MFCD09037341
UN Number NONH for all modes of transport
Synonym (5 S)-5-Methyl-5-phenyl-3-(phenylamino)-2,4-imidazolidinedione; Fenamidone Metabolite
Formula Weight 281.31
Grade Analytical Standard
Physical Form Neat
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH