missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, MilliporeSigma™ Supelco™

Catalog No. 111019112
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019112 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019112 Supplier Merck Emd Millipore Supplier No. 33674100MGR
Only null left

Acetamiprid, is a neonicotinoid insecticide, which finds applications in agriculture for pest control. It can also act as acetylcholine nicotinic receptor.

Agriculture

Specifications

Product Line PESTANAL
CAS 160430-64-8
Physical Form Neat
Linear Formula C10H11 ClN4
MDL Number MFCD06201842
Melting Point 100°C to 102°C
Synonym Acetamiprid
Type Analytical Standard
Quantity 100 mg
Formula Weight 222.67
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH