missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Flumorph, MilliporeSigma™ Supelco™

Catalog No. 111013531
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013531 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013531 Supplier Merck Emd Millipore Supplier No. 3703825MG
Only null left

Flumorph is a oomycete carboxylic acid amide (CAA) fungicide, which shows strong activity against Peronospora and Phytophthora diseases in vegetables and fruits.

Agriculture

Specifications

Product Line PESTANAL
CAS 211867-47-9
Physical Form Neat
Linear Formula C21H22 FNO4
Synonym (EZ)-4-[3-(3,4-Dimethoxyphenyl)-3-(4-fluorophenyl)acryloyl]morpholine
Type Analytical Standard
Quantity 25 mg
Formula Weight 371.40
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH