missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Ethofumesate-2-keto, MilliporeSigma™ Supelco™

Catalog No. 111018229
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018229 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018229 Supplier Merck Emd Millipore Supplier No. 3388810MGR
Only null left

Ethofumesate-2-keto is a metabolite of the herbicide, ethofumesate used for the control of weeds in strawberry and beet crops. It inhibits the growth of meristems, retards cellular division and limits the formation of cuticules.

Agriculture

Specifications

Product Line PESTANAL
CAS 26244-33-7
Physical Form Neat
Linear Formula C11H12O5 S
MDL Number MFCD01632378
Synonym 3,3-Dimethyl-2-oxo-2,3-dihydrobenzofurane-5-yl methane sulfonate; Methane sulfonic acid 3,3-dimethyl-2-oxo-2,3-dihydrobenzofuran-5-yl ester
Type Analytical Standard
Quantity 10 mg
Formula Weight 256.27
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH