missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Ethalfluralin, MilliporeSigma™ Supelco™

Catalog No. 111019985
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019985 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019985 Supplier Merck Emd Millipore Supplier No. 45472250MG
Only null left

Ethalfluralin belongs to the group of dinitroaniline herbicides used as a pre-emergence herbicide for controlling weeds in a variety of crops such as potatoes, pumpkins, squash, canola, dry peas, drybeans, soybeans, sunflowers, peanuts, etc.

Agriculture

Specifications

Product Line PESTANAL
CAS 55283-68-6
Physical Form Neat
Linear Formula C13H14F3N3O4
MDL Number MFCD00078681
Type Analytical Standard
Quantity 250 mg
Formula Weight 333.26
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH