missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Diazinon Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111020152
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020152 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020152 Supplier Merck Emd Millipore Supplier No. 458421ML
Only null left

Diazinon, belongs to the organophosphorus class of pesticides, extensively used in agriculture. Its mode of action involves the inhibition of cholinesterase in insects and mammals and also brings about non-reversible phosphorylation of esterases in the central nervous system of the organism.

Agriculture and Environmental

Specifications

Product Line PESTANAL
CAS 333-41-5
Flash Point 2°C
Linear Formula C12H21N2O3 PS
MDL Number MFCD00036204
Type Analytical Standard
Quantity 1 mL
Formula Weight 304.35
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH