missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Desnitro-imidacloprid hydrochloride, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 127202-53-3 |
| Physical Form | Neat |
| Linear Formula | C9H11 ClN4 · HCl |
| MDL Number | MFCD22373625 |
| Synonym | 1-(2-Chloro-pyridin-5-yl-methyl)-2-imino-imidazolidine hydrochloride |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 247.12 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH