missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cycloate, MilliporeSigma™ Supelco™

Catalog No. 111020022
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020022 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020022 Supplier Merck Emd Millipore Supplier No. 45408250MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 1134-23-2
Flash Point >100°C
Physical Form Neat
Linear Formula C11H21 NOS
MDL Number MFCD00055341
Synonym S-Ethyl N-cyclohexyl-N-ethylthiocarbamate
Type Analytical Standard
Quantity 250 mg
Formula Weight 215.36
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH