missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Chlorflurenol, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 2464-37-1 |
| Physical Form | Neat |
| Linear Formula | C14H9 ClO3 |
| MDL Number | MFCD00056359 |
| Synonym | 2-Chloro-9-hydroxyflurene-9-carboxylic acid |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 260.67 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH