missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Chinomethionate, MilliporeSigma™ Supelco™

Catalog No. 111020195
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020195 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020195 Supplier Merck Emd Millipore Supplier No. 45372250MG
Only null left
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line PESTANAL
CAS 2439-01-2
Physical Form Neat
Linear Formula C10H6N2 OS2
MDL Number MFCD00055387
Synonym Oxythioquinox
Type Analytical Standard
Quantity 250 mg
Formula Weight 234.30
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH