missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Carbendazim-d3, MilliporeSigma™ Supelco™

Catalog No. 111018238
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018238 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018238 Supplier Merck Emd Millipore Supplier No. 3241310MG
Only null left
Agriculture

Specifications

Product Line PESTANAL
CAS 1255507-88-0
Physical Form Neat
Linear Formula C9D3H6N3O2
MDL Number MFCD17015247
Synonym Methyl-d3 2-benzimidazolecarbamate
Type Analytical Standard
Quantity 10 mg
Formula Weight 194.21
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH