missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Bensultap Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 17606-31-4 |
| Molecular Formula | C17H21NO4S4 |
| Linear Formula | C17H21NO4S4 |
| Quantity | 100 mg |
| MDL Number | MFCD00210246 |
| UN Number | UN 3077 9 / PGIII |
| Synonym | S,S'-[2-(Dimethylamino)trimethylene] di(benzenethiosulfonate) |
| Formula Weight | 431.61 |
| Grade | Analytical Standard |
| Physical Form | Neat |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH