missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Acetamiprid-N-desmethyl, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 190604-92-3 |
| Physical Form | Neat |
| Linear Formula | C9H9 ClN4 |
| MDL Number | MFCD08690484 |
| Synonym | N-(6-Chloro-3-pyridylmethyl)-N′-cyano-acetamidine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 208.65 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH