missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 3-(Methylphosphinico)propionic acid, MilliporeSigma™ Supelco™

Catalog No. 111013198
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013198 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013198 Supplier Merck Emd Millipore Supplier No. 31264100MG

3-(Methylphosphinico)propionic acid is a metabolite of the pesticide, glufosinate ammonium.

Agriculture

Specifications

CAS 15090-23-0
Quantity 100 mg
Formula Weight 152.09
Physical Form Neat
Product Line PESTANAL

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH