missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 3-Chlorotoluene Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022468
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022468 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022468 Supplier Merck Emd Millipore Supplier No. 366961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3-Chlorotoluene is a chlorinated solvent, and a major environmental pollutant. It is primarily used in textile, peroxide, pesticide, pharmaceutical, and dye industries.

Specifications

Melting Point -48°C (literature)
CAS 108-41-8
Density 1.072 g/mL (at 25°C (literature))
Quantity 1 g
Flash Point 52°C (Closed Cup)
Physical Form Neat
Refractive Index n20/D 1.522 (literature)
Molecular Formula C7H7Cl
Linear Formula CH3C6H4Cl
MDL Number MFCD00000595
UN Number UN 2238 3 / PGIII
Boiling Point 160°C to 162°C (literature)
Formula Weight 126.58
Grade Analytical
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH