missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ 2,4-D butylglycol ester, MilliporeSigma™ Supelco™
Description
Agriculture
Specifications
Specifications
| Product Line | PESTANAL |
| CAS | 1929-73-3 |
| Physical Form | Neat |
| Linear Formula | C14H18 Cl2O4 |
| MDL Number | MFCD00126849 |
| Synonym | 2,4-D butylglycol ester; 2,4-Dichlorophenoxyacetic acid butylglycol ester; 2-Butoxyethyl 2,4-dichlorophenoxyacetate |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 321.20 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH