missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic acid, 75-80% aq. soln.

Catalog No. AA1141314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1141314 25 g
AA1141306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1141314 Supplier Thermo Scientific Chemicals Supplier No. 01141314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13768-11-1 | HO4Re | 251.21 g/mol

Perrhenic acid is used in the manufacturing of superconducting synthetic metals and X-ray targets. It is used as a hydrocracking catalyst in petroleum industry and used for the dehydrogenation process and epoxidation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13768-11-1
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
Synonym perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
PubChem CID 83718
IUPAC Name hydroxy(trioxo)rhenium
SMILES O[Re](=O)(=O)=O

Specifications

Color Colorless through Yellow
Physical Form Liquid
Quantity 25 g
Appearance Colorless to pale yellow liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 75-80%
Density 2.42
Solubility Information Soluble in water.
Formula Weight 251.26
Concentration or Composition (by Analyte or Components) Perrhenic acid: 80.0%; Water: 20.0%
Chemical Name or Material Perrhenic acid
Show More Show Less
Hazard Category H302-H314
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: I
EINECSNumber 237-380-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only