missing translation for 'onlineSavingsMsg'
Learn More

Perphenazine 97.0+%, TCI America™

Catalog No. p-7135339
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19705G 5 g
P197025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19705G Supplier TCI America Supplier No. P19705G
Only null left

Chemical Identifiers

CAS 58-39-9
Molecular Formula C21H26ClN3OS
Molecular Weight (g/mol) 403.97
MDL Number MFCD00056798
InChI Key RGCVKNLCSQQDEP-UHFFFAOYSA-N
Synonym 2-Chloro-10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazine, 4-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-piperazineethanol
PubChem CID 4748
ChEBI CHEBI:8028
IUPAC Name 2-{4-[3-(2-chloro-10H-phenothiazin-10-yl)propyl]piperazin-1-yl}ethan-1-ol
SMILES OCCN1CCN(CCCN2C3=CC=CC=C3SC3=CC=C(Cl)C=C23)CC1

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Formula Weight 403.97
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Perphenazine
EINECSNumber (9)-0456
RTECSNumber TL7175000
TSCA No
Recommended Storage Refrigerator